Gaskets for SVL threads, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H979-32 H979-24 H979-12
BOHLH979-32EA 10.4 EUR
BOHLH979-32 BOHLH979-24 BOHLH979-12
Gaskets for SVL threads, BOLA
Afdichtingen Pakkingen
Gasket, made of PTFE/Silicone, for tubing, tubes or probes inserted through Torion threads.
  • Silicone ring with double sided PTFE
  • Suitable for Torion / SVL threaded hoses
  • Temperature resistance: −200 to +250 °C