Screw caps HT (high temperature), BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H994-14 H994-18 H994-32 H994-25 H994-45
BOHLH994-14EA 4.2 EUR
BOHLH994-14 BOHLH994-18 BOHLH994-32 BOHLH994-25 BOHLH994-45
Screw caps HT (high temperature), BOLA
Sluitingen Flesdoppen
Screw caps, made of PPS, with handy knurl and integrated PTFE membrane gasket.
  • Very good chemical resistance
  • Temperature resistant from −20 to +250 °C
  • Suitable for vaccum application
Suitable for bottles and vessels with GL threads.
Certificaten: Gasket is safe according to VDI/VDE guideline 2480 and fulfils FDA 21 CFR 177-1550 requirements.