Non return valves, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E680-23 E680-27 E680-31 E680-21
BOHLE680-23EA 477 EUR
BOHLE680-23 BOHLE680-27 BOHLE680-31 BOHLE680-21
Non return valves, BOLA
Ventielen Terugslagventielen
These non return valves are made from PTFE, with nuts made from glass fibre reinforced PTFE for connecting tubing or tubes.
  • Universally chemically resistant
  • Flow medium comes into contact with PTFE or PFA only
The built-in lock function only allows flow in one direction, the flow direction is marked by an arrow.