Reducing tubing connectors, straight, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D572-02 D572-04 D572-06
BOHLD572-02EA 13.2 EUR
BOHLD572-02 BOHLD572-04 BOHLD572-06
Reducing tubing connectors, straight, BOLA
Fittingen en connectoren
Straight fitting, made of PTFE, with two connectors for elastic tubing with different inner diameters.
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Vacuum: suitable