Tubing connectors, Y-shaped, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D576-06 D576-02 D576-04 D576-08
BOHLD576-06EA 67.1 EUR
BOHLD576-06 BOHLD576-02 BOHLD576-04 BOHLD576-08
Tubing connectors, Y-shaped, BOLA
Fittingen en connectoren
Connectors for tubing, made of PTFE, Y-shape.
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Suitable for vacuum