Lithium hydroxide, anhydrous 98%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
379181000 379185000 379180025
ACRO379181000EA 78.2 EUR
ACRO379181000 ACRO379185000 ACRO379180025
Lithium hydroxide, anhydrous 98%, pure
Lithium hydroxide
Formule: LiOH
MW: 23.95 g/mol
Kookpunt: 924
Smeltpunt: 462 °C
Dichtheid: 1.43 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011095
CAS nummer: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8 II
Merck Index: 13,05556
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Titration with HCl ≥97.5 %