Disodium hydrogen phosphate, Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
567550-1 567550-500
567550-1EA 192 EUR
567550-1 567550-500
Disodium hydrogen phosphate, Millipore®
Disodium hydrogen phosphate
Formule: Na₂HPO₄
MW: 141.96 g/mol
Smeltpunt: 250
Dichtheid: 1.53 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003496
CAS nummer: 7558-79-4
EINECS: 231-448-7
Bereik: 01-2119489797-11
Merck Index: 12,08805