Potassium perrhenate (64% Re) ≥99% (metals basis)
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 99%
CAS No.: 10466-65-6
Molecular Formula: KReO4
Formula Weight: 289.30
Physical Form: Crystalline
Density: 4.887
Refractive Index: 1.643
Appearance: White
Odour: Odourless
Melting Point: 550°
Boiling Point: 1360-1370°
UN No.: 1479
MDL No.: MFCD00011365
A powerful oxidizer
Formule:
KO₄Re Kookpunt: 1360…1370 °C Smeltpunt: 550 °C Dichtheid: 4,887 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011365 CAS nummer: 10466-65-6 EINECS: 233-953-8 UN: 1479 ADR: 5.1,II |