Natriumsulfaat ≥99.9955% (metals basis), Puratronic®
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Purity: 99.9955%
CAS No.: 7757-82-6
Molecular Formula: Na2SO4
Formula Weight: 142.04
Physical Form: Powder
Density: 2.680
Refractive Index: 1.484
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 884°
Merck Reference: 14,8680
MDL No.: MFCD00003504
Formule:
Na₂SO₄ Kookpunt: 1429 °C (1013 hPa) Smeltpunt: 884 °C Dichtheid: 2,680 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003504 CAS nummer: 7757-82-6 EINECS: 231-820-9 Merck Index: 14,8680 |