Micro pipettes, end-to-end
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2911109 2911110
MARI2911109EA 0 EUR
MARI2911109 MARI2911110
Micro pipettes, end-to-end
Pipetten Micro-capillairen
These micro pipettes made of glass can dispense more accurately than conventional pipettes. They are used for dispensing very small amounts of liquids.
  • For single use
  • Sodium-heparinized (80 iu/ml)
Certificaten: Compliance with DIN ISO 7550. Approved for in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark.
Verpakking: 100 pieces in a colour coded dispenser vial, 10 vials in a box.
Order Now
Specificaties
Environmentally Preferable