Centrifuge tubes
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3906001 3933041
MARI3906001EA 84.8 EUR
MARI3906001 MARI3933041
Centrifuge tubes
Buisjes Centrifugebuisjes
Borosilicate glass 3,3.
  • Heavy walled
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Short conical or round bottom.