Capillaire kolommen, BPX1
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054800 054801 054802 054803 054777 0548025
568-0456EA 0 EUR
568-0456 568-0457 568-0458 568-0460 568-0452 568-0459
Capillaire kolommen, BPX1
Kolommen voor chromatografie
These columns are specifically designed for high temperature hydrocarbon analysis. Ideal for simulated distillation.
100% dimethyl polysiloxane
Max. temperature: Polyimide 0,1 - 0,9 µm film thickness 30 to 400 °C; 2,65 µm film thickness 30 to 370 °C aluminium 30 to 430 °C
Suitable replacement for: DB-2887, DB-HT Sim Dis, HP-1, Petrocol 2887, Petrocol EX2887, Rtx-2887