Capillaire kolommen, BPX35
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054701 054702 054711 0547158 054724 054737 054699
558-0250EA 672 EUR
558-0250 568-0420 568-0426 568-0430 568-0437 568-0442 568-0418
Capillaire kolommen, BPX35
Kolommen voor chromatografie
These columns are suitable for environmental analyses, pesticides/herbicides, drugs of abuse, pharmaceuticals, polynuclear aromatic hydrocarbons and GC-MS applications.
35% phenyl polysilphenylene-siloxane
Max. temperature: 10 to 330/360 °C
Suitable replacement for: DB-35, DB-35ms, Rtx-35, HP-35, HP-35MS, SPB-35, MDN-35.