Culture and test tubes with screw caps
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3737737 3737852 3737862
MARI3737737EA 198 EUR
MARI3737737 MARI3737852 MARI3737862
Culture and test tubes with screw caps
Buisjes Reageerbuisjes
Borosilicate glass, clear, with black screw cap.
  • With marking spot
  • Good chemical resistance
  • Good temperature resistance