SGE GC Capillary Column, CYDEX-B
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054900 054901 054950
568-0464EA 806 EUR
568-0464 568-0465 568-0241
SGE GC Capillary Column, CYDEX-B
Kolommen voor chromatografie
The SGE CYDEX-B GC Capillary Column provides a great low-polarity option for GC analyses. Recommended for separation of enantiomeric compounds found in natural products.
  • Permethylated Beta-Cyclodextrin phase for the separation of chiral compounds
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.