Capillaire kolommen, BP21
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054468 054469 054474 054465 054466 054471 054472 054477 054478 054480
568-0405EA 772 EUR
568-0405 568-0406 568-0411 568-0402 568-0403 568-0408 568-0409 568-0413 568-0414 568-0415
Capillaire kolommen, BP21
Kolommen voor chromatografie
These columns are suitable for volatile free acids, fatty acid methyl esters, alcohols, aldehydes, acrylates, ketones.
Polyethylene glycol (PEG) - TPA treated
Max. temperature: 35 to 240/250 °C
Suitable replacement for: DB-FFAP, HP-FFAP, Stabilwax-DA, CPWax-58CB