Calciumchloride, ultra dry ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Chloro calcium, Calcium dichloride, CaCl2
Ampuled under argon
35685.04 35685.09
35685.04EA 103 EUR
35685.04 35685.09
Calciumchloride, ultra dry ≥99.9% (metals basis)
Calciumchloride
Purity: 99.9% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.15 Appearance: White Melting Point: 782° Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903
Formule: CaCl₂
MW: 110.98 g/mol
Kookpunt: >1600 °C
Smeltpunt: 782 °C
Dichtheid: 2.15
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010903
CAS nummer: 10043-52-4
EINECS: 233-140-8
Bereik: 01-2119494219-28
Merck Index: 14,1659