Ammonium sulphite monohydrate ≥92%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
17996.A1
17996.A7
17996.A1EA
52.1
EUR
17996.A1
17996.A7
Ammonium sulphite monohydrate ≥92%
Ammonium sulphite monohydrate
Purity: 92% min
CAS No.: 7783-11-1
Molecular Formula: H8N2O3S.H2O
Formula Weight: 134.15 (116.14anhy)
Physical Form: Crystalline
Density: 1.41
Refractive Index: 1.5150
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 60-70° dec.
Merck Reference: 14,557
MDL No.: MFCD00066094
Formule:
H₈N₂O₃S·1H₂O MW: 134.16 g/mol Smeltpunt: 60…70 °C (ontbinding) Dichtheid: 1.41 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00066094 CAS nummer: 7783-11-1 EINECS: 233-484-9 |