Palladium 5% on carbon, dry for use as a catalyst Type 87L
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A12623.09 A12623.18
A12623.09EA 178 EUR
A12623.09 A12623.18
Palladium 5% on carbon, dry for use as a catalyst Type 87L
Palladium
CAS No.: 7440-05-3 Molecular Formula: Pd Merck Reference: 14,6989 UN No.: 1325 MDL No.: MFCD00011167
Formule: Pd
MW: 106.420 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
UN: 1325
ADR: 4.1 III
Merck Index: 14,6989