Phenoxyacetamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A14934.06 A14934.14
A14934.06EA 33 EUR
A14934.06 A14934.14
Phenoxyacetamide ≥98%
Phenoxyacetamide

Purity: 98% CAS No.: 621-88-5 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 100-104° Beilstein Reference: 1941781

Formule: C₈H₉NO₂
Smeltpunt: 100…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00017140
CAS nummer: 621-88-5
EINECS: 210-713-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR