Ethylphosphonic acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B22559.03
B22559.06
B22559.14
AB133491.0025
B22559.03EA
37.6
EUR
B22559.03
B22559.06
B22559.14
ABCRAB133491.0025
Ethylphosphonic acid ≥98%
Ethylphosphonic acid
Purity: 98+%
CAS No.: 6779-09-5
Molecular Formula: C2H7O3P
Formula Weight: 110.06
Melting Point: 58-62°
Beilstein Reference: 1743032
UN No.: 3261
MDL No.: MFCD00002141
Formule:
C₂H₇O₃P Smeltpunt: 58…62 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002141 CAS nummer: 6779-09-5 EINECS: 229-839-2 UN: 3261 ADR: 8,II |