1-Butyl-2-urea 96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
L02854.22 L02854.36
L02854.22EA 154 EUR
L02854.22 L02854.36
1-Butyl-2-urea 96%
1-Butyl-2-urea
Purity: 96% CAS No.: 592-31-4 Molecular Formula: CH3(CH2)3NHCONH2 Formula Weight: 116.16 Melting Point: 92-96° Beilstein Reference: 1744775
Formule: C₅H₁₂N₂O
Smeltpunt: 92…96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007956
CAS nummer: 592-31-4
EINECS: 209-748-4
Order Now
Specificaties
Environmentally Preferable