L(+)-α-Phenylglycine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-Phg-OH, L-(+)-2-Phenylglycine

4031152.0025 4031152.0100 4031152.0500
BACE4031152.0025EA 0 EUR
BACE4031152.0025 BACE4031152.0100 BACE4031152.0500
L(+)-α-Phenylglycine
L(+)-α-Phenylglycine
Formule: C₈H₉NO₂
MW: 151.16 g/mol
MDL Nummer: MFCD00064403
CAS nummer: 2935-35-5
EINECS: 220-909-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR