D(-)-α-Phenylglycine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4031153.0100 4031153.0500
BACE4031153.0100EA 0 EUR
BACE4031153.0100 BACE4031153.0500
D(-)-α-Phenylglycine
D-(-)-α-Phenylglycine
Formule: C₈H₉NO₂
MDL Nummer: MFCD00008061
CAS nummer: 875-74-1
EINECS: 212-876-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR