Aluminiumchloride, anhydrous 99.999% (trace metals basis), powder for analysis
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aluminum chloride
364810200 364810100 364811000
ACRO364810200EA 181 EUR
ACRO364810200 ACRO364810100 ACRO364811000
Aluminiumchloride, anhydrous 99.999% (trace metals basis), powder for analysis
Aluminiumchloride
Formule: AlCl₃
MW: 133.34 g/mol
Kookpunt: 180
Smeltpunt: 194 °C
Dichtheid: 2.44 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003422
CAS nummer: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤0.001 %
Iron (Fe) ≤5 ppm