2-Mercaptonicotinic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Thionicotinic acid

SB00659.1 SB00659.10 SB00659.50
MAYBSB00659.1EA 0 EUR
MAYBSB00659.1 MAYBSB00659.10 MAYBSB00659.50
2-Mercaptonicotinic acid
2-Mercaptonicotinic acid
Formule: C₆H₅NO₂S
MDL Nummer: MFCD00010102
CAS nummer: 38521-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR