Benzylthiourea

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00565.1 TL00565.10 TL00565.50
MAYBTL00565.1EA 0 EUR
MAYBTL00565.1 MAYBTL00565.10 MAYBTL00565.50
Benzylthiourea
Benzylthiourea
Formule: C₈H₁₀N₂S
MDL Nummer: MFCD00041370
CAS nummer: 621-83-0
EINECS: 210-709-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR