Aluminiumchloride, ultra dry ≥99.999% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Aluminum chloride
Ampuled under argon
44313.14
44313.22
44313.14EA
267
EUR
44313.14
44313.22
Aluminiumchloride, ultra dry ≥99.999% (metals basis)
Aluminiumchloride
Purity: 99.999%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Powder, Ampouled under argon
Density: 2.44
pH: 2,4
Melting Point: 190°subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Formule:
AlCl₃ Kookpunt: 180 °C (1013 hPa) Dichtheid: 2,44 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003422 CAS nummer: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |