Ammonium sulfite 35% in aqueous solution, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.01221.1000 8.01221.2500 8.01221.9025 8.01221.9200
8.01221.1000EA 20.3 EUR
8.01221.1000 8.01221.2500 8.01221.9025 8.01221.9200
Ammonium sulfite 35% in aqueous solution, Sigma-Aldrich®
Ammonium sulphite
Formule: H₈N₂O₃S
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 10196-04-0
EINECS: 233-484-9
Order Now
Specification Test Results
Solubility (20°C) Soluble
Melting Point 4 - 8 °C
Boiling Point 101 °C
Density 1.19 g/cm3 (20 °C)
pH 7.5 - 7.6 (H2O, 20 °C)