Cysteaminium chloride for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-MEA
M-Clarity™ quality level = MQ200
8.02835.0025 8.02835.0100 8.02835.0010 8.02835.1001
8.02835.0025EA 54.4 EUR
8.02835.0025 8.02835.0100 8.02835.0010 8.02835.1001
Cysteaminium chloride for synthesis, Sigma-Aldrich®
Cysteaminium chloride
Formule: C₂H₇NS·HCl
MW: 113.61 g/mol
Smeltpunt: 67…71 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012904
CAS nummer: 156-57-0
EINECS: 205-858-1
Order Now
Specification Test Results
Assay (iodometric) ≥ 97.0 %
Melting range (lower value) ≥ 61 °C
Melting range (upper value) ≤ 67 °C
Identity (IR) passes test