3-Oxoglutaric acid
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid
31350.02
31350.03
SERA31350.02EA
0
EUR
SERA31350.02
SERA31350.03
3-Oxoglutaric acid
3-Oxoglutaric acid
Formule:
C₅H₆O₅ MW: 146.1 g/mol Kookpunt: 323 Smeltpunt: 138 Dichtheid: 1.55 g/cm³ (20 °C) Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00002711 CAS nummer: 542-05-2 EINECS: 208-797-9 Merck Index: 13,00069 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...