Indium(I) chloride, anhydrous ≥99.995% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Packaged under argon.
40033.02
40033.06
40033.14
40033.02EA
24.5
EUR
40033.02
40033.06
40033.14
Indium(I) chloride, anhydrous ≥99.995% (metals basis)
Indium(I) chloride
Purity: 99.995%
CAS No.: 13465-10-6
Molecular Formula: InCl
Formula Weight: 150.27
Physical Form: Crystalline, Packaged under argon
Density: 4.19
Melting Point: ca 225°
Boiling Point: 608°
UN No.: 3260
MDL No.: MFCD00148875
Formule:
InCl Kookpunt: 608 °C Smeltpunt: ∼225 °C Dichtheid: 4,19 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00148875 CAS nummer: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |