(±)-Methyl-2-bromobutyrate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Methyl 2-bromobutyrate
B1462.0500
B1462-25G
B1462-500G
TCIAB1462.0500EA
0
EUR
TCIAB1462.0500
TCIAB1462-25G
TCIAB1462-500G
(±)-Methyl-2-bromobutyrate ≥97.0% (by GC)
(±)-Methyl-2-bromobutyrate
Formule:
C₅H₉BrO₂ MW: 181.03 g/mol Kookpunt: 172°C Vlampunt: 68°C |
MDL Nummer:
MFCD00009666 CAS nummer: 3196-15-4 EINECS: 221-699-0 UN: 3265 ADR: 8 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...