Accucore™ kits
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17126-0521303V 17126-1021303V 17126-1521303V 17X260521303VA 17X261021303VC 17X261521303VC
554-1960EA 1870 EUR
554-1960 554-1961 554-1962 554-1963 554-1970 554-1971
Accucore™ kits
Kolommen voor chromatografie
Accucore™ kits validate the reproducibility of Accucore™ HPLC columns, or verify which Accucore™ HPLC column offers the best separation.
Accucore™ validation kit: Validate the reproducibility of Accucore™ HPLC columns.
Accucore™ narrow selectivity kit: Verify which Accucore™ HPLC column offers the best separation over a narrow selectivity range.
Accucore™ wide selectivity kit: Verify which Accucore™ HPLC column offers the best separation over a wide selectivity range.
Accucore™ polar selectivity kit: Verify which Accucore™ HPLC column offers the best separation for polar analytes.
Leveringsinformatie: The Accucore™ validation kit contains three Accucore™ C18 columns. The Accucore™ narrow selectivity kit contains one each of Accucore™ C18, RP-MS and aQ columns. The Accucore™ wide selectivity kit contains one each of Accucore™ C18, Phenyl-Hexyl and PFP columns. The Accucore™ polar selectivity kit contains one each of Accucore™ aQ, PFP and HILIC columns.