Boorzuur 99.99% (trace metals basis), extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
315181000 315180025 315185000
ACRO315181000EA 74.6 EUR
ACRO315181000 ACRO315180025 ACRO315185000
Boorzuur 99.99% (trace metals basis), extra pure
Boorzuur
Formule: B(OH)₃
MW: 61.83 g/mol
Kookpunt: 185
Smeltpunt: 169 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011337
CAS nummer: 10043-35-3
EINECS: 233-139-2
Bereik: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Total trace metal impurities ≤100 ppm
Titration with NaOH ≥98.5 %