Kiezelgel 3-6 mm, non toxic for drying
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
357400010 357400050 357400250
ACRO357400010EA 59.1 EUR
ACRO357400010 ACRO357400050 ACRO357400250
Kiezelgel 3-6 mm, non toxic for drying
Kiezelgel
Formule: SiO₂
Kookpunt: 2230 °C (1013 hPa)
Smeltpunt: 1710 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 7631-86-9
Bereik: 01-2119379499-16
Order Now
Specification Test Results
Appearance Colorless to white Irregular granules
Particle size 3.0 to 6.0 mm
Residual water ≤2.0 % (140°C, 4 h)
Water adsorption ≥20.0 % (40% rH)