Kiezelgel, Pore size: 60 Å

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0712.1 0712.2
ROTH0712.1EA 0 EUR
ROTH0712.1 ROTH0712.2
Kiezelgel, Pore size: 60 Å
Kiezelgel
Formule: SiO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR