Kiezelgel, Pore size: 60 Å

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P091.1 P091.2 P091.3
ROTHP091.1EA 43.9 EUR
ROTHP091.1 ROTHP091.2 ROTHP091.3
Kiezelgel, Pore size: 60 Å
Kiezelgel
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: 2230
Smeltpunt: 1710
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR