Kiezelgel, Pore size: 60 Å 70-230 mesh, high purity for column chromatography, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
60741-1KG 60741-25KG 60741-5KG
60741-1KGEA 122 EUR
60741-1KG 60741-25KG 60741-5KG
Kiezelgel, Pore size: 60 Å 70-230 mesh, high purity for column chromatography, Supelco®
Kiezelgel
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: 2230
Smeltpunt: 1710
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=269b27dcefc3b5c7fb165351f35e8d8c78e597a9&vid=b12160b7bd956d4812c0c9d911a65dbdcfeefa50&prid=web&clid=SAPDAM