Tin selenide ≥99.999% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

18781.06 18781.14
18781.06EA 123 EUR
18781.06 18781.14
Tin selenide ≥99.999% (metals basis)
Tin selenide

Purity: 99.999% CAS No.: 1315-06-6 Molecular Formula: SnSe Formula Weight: 197.65 Physical Form: 3-12mm Pieces Melting Point: 861° UN No.: 3077 MDL No.: MFCD00054049

Formule: SeSn
Smeltpunt: 861 °C
Dichtheid: 6,18 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054049
CAS nummer: 1315-06-6
EINECS: 215-257-6
UN: 3077
ADR: 9,III
Merck Index: 14,8789

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR