Bismuth(III) chloride 99.999% (trace metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Bismuth(III) chloride, Bismuth trichloride
318350050 318350250
ACRO318350050EA 69 EUR
ACRO318350050 ACRO318350250
Bismuth(III) chloride 99.999% (trace metals basis)
Bismuth(III) chloride
Formule: BiCl₃
MW: 315.34 g/mol
Kookpunt: 447 °C (760 mmHg)
Smeltpunt: 230…232 °C
Dichtheid: 4.75 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003461
CAS nummer: 7787-60-2
EINECS: 232-123-2
Merck Index: 13,01261
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Total trace metal impurities ≤10 ppm