Tellurium tetrachloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
194740250 194741000
ACRO194740250EA 114 EUR
ACRO194740250 ACRO194741000
Tellurium tetrachloride 99%
Tellurium tetrachloride
Formule: Cl₄Te
Kookpunt: 380 °C (760 mmHg)
Smeltpunt: 224 °C
Dichtheid: 3,26 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011262
CAS nummer: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8,II
Merck Index: 12,09279
Order Now
Specification Test Results
Appearance Off-white to light yellow Powder
Titration Argentometric ≥98.5 % (ex Cl)