Silver chloride ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
11421.14
11421.22
11421.36
11421.69
11421.14EA
139
EUR
11421.14
11421.22
11421.36
11421.69
Silver chloride ≥99.9% (metals basis)
Silver chloride
Purity: 99.9%
CAS No.: 7783-90-6
Molecular Formula: AgCl
Formula Weight: 143.32
Physical Form: Crystalline
Density: 5.56
Refractive Index: 2.071
Sensitivity: Light Sensitive
Appearance: White
Odour: Odourless
Melting Point: 455°
Boiling Point: 1550°
Merck Reference: 14,8509
UN No.: 3077
MDL No.: MFCD00003399
Formule:
AgCl Kookpunt: 1550 °C Smeltpunt: 455 °C Dichtheid: 5,56 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003399 CAS nummer: 7783-90-6 EINECS: 232-033-3 UN: 3077 ADR: 9,III Merck Index: 14,8509 |