Silver chloride, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ300
1.19203.0025 1.19203.0100 1.19203.1000
1.19203.0025EA 479 EUR
1.19203.0025 1.19203.0100 1.19203.1000
Silver chloride, Supelco®
Silver chloride
Formule: AgCl
Kookpunt: 1554 °C (1013 hPa)
Smeltpunt: 455 °C
Dichtheid: 5,56 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003399
CAS nummer: 7783-90-6
EINECS: 232-033-3
UN: 3077
ADR: 9,
Merck Index: 13,08582
Order Now
Specification Test Results
Solubility 0.00188 g/l (25 °C)
Melting Point 455 °C
Molar Mass 143.32 g/mol
Boiling Point 1554 °C (1013 hPa)
Density 5.56 g/cm3 (20 °C)
Specificaties
Environmentally Preferable