Silver(I) oxide, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ300
1.19208.0025 1.19208.0100
1.19208.0025EA 203 EUR
1.19208.0025 1.19208.0100
Silver(I) oxide, Supelco®
Silver(I) oxide
Formule: Ag₂O
Smeltpunt: 800…830 °C
Dichtheid: 7,14 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003404
CAS nummer: 20667-12-3
EINECS: 243-957-1
UN: 3085
ADR: 5.1 (8),
Merck Index: 12,08665
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 200 °C decomposes
Molar Mass 231.74 g/mol
Bulk Density 950 kg/m3
Density 7.2 g/cm3 (20 °C)