Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Zirconium dioxide, Zirconium Oxide
36319.18 36319.30
36319.18EA 98.4 EUR
36319.18 36319.30
Zirconium(IV) oxide ≥99.7% (metals basis excluding Hf) (max. 75ppm Hf)
Zirconium(IV) oxide
Purity: 99.7% CAS No.: 1314-23-4 Molecular Formula: ZrO2 Formula Weight: 123.22 Physical Form: 3-12mm Sintered Lump Density: 5.89 Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310
Formule: ZrO₂
MW: 123.22 g/mol
Kookpunt: ∼5000 °C
Smeltpunt: ∼2700 °C
Dichtheid: 5.89
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180