1-Chloronaphthalene for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Triisodecyl phosphite, α-Chloronaphthalene
M-Clarity™ quality level = MQ200
8.20315.1000 8.20315.0100 8.20315.0025
8.20315.1000EA 96.7 EUR
8.20315.1000 8.20315.0100 8.20315.0025
1-Chloronaphthalene for synthesis, Sigma-Aldrich®
1-Chloronaphthalene
Formule: C₁₀H₇Cl
MW: 162.62 g/mol
Kookpunt: 263
Smeltpunt: –2.5 °C
Dichtheid: 1.1938 g/cm³ (20 °C)
Vlampunt: 121
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003874
CAS nummer: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: 9 III
Merck Index: 13,02168
Order Now
Specification Test Results
Solubility 0.009 g/l (20 °C)
Melting Point -6 °C
Molar Mass 162.61 g/mol
Boiling Point 259 °C (1013 hPa)
Vapor Pressure 0.05 hPa (20 °C)
Flash Point 115 °C
Density 1.19 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) (undiluted)
Ignition Point 500 °C
Synonyms α-Naphthyl chloride
Hill Formula C10H7Cl