2-Methoxy-1-nitronaphthalene
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NRB01117.250
NRB01117.1
MAYBNRB01117.250EA
0
EUR
MAYBNRB01117.250
MAYBNRB01117.1
2-Methoxy-1-nitronaphthalene
2-Methoxy-1-nitronaphthalene
Formule:
C₁₁H₉NO₃ MW: 203.2 g/mol |
MDL Nummer:
MFCD00100119 CAS nummer: 4900-66-7 EINECS: 225-529-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...