1-Naftol zS, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
MQ200
8.22289.0250 8.22289.1000 8.22289.9050
8.22289.0250EA 46.5 EUR
8.22289.0250 8.22289.1000 8.22289.9050
1-Naftol zS, Sigma-Aldrich®
1-Naftol
Formule: C₁₀H₈O
MW: 144.17 g/mol
Kookpunt: 282
Smeltpunt: 94
Dichtheid: 1.224 g/cm³ (20 °C)
Vlampunt: 125
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003930
CAS nummer: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,06409
Order Now
Specification Test Results
Solubility 0.1 g/l (20 °C)
Melting Point 94 - 96 °C
Molar Mass 144.17 g/mol
Bulk Density 450 kg/m3
Boiling Point 288 °C (1013 hPa)
Vapor Pressure 1.3 hPa (94 °C)
Flash Point 125 °C
Density 1.28 g/cm3 (20 °C)
Ignition Point 510 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials