Glass lined tubing, GLT™
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
082707 082712 082717 082718 082719 082720 082724 082725 082734 0827352 0827353 082737 0827375 082738 082740 082742 082745 082747 082760 082767 082768 082770 082708 082710 08277028 0827075 0827120 0827615
SGEA082707EA 42.7 EUR
SGEA082707 SGEA082712 SGEA082717 SGEA082718 SGEA082719 SGEA082720 SGEA082724 SGEA082725 SGEA082734 SGEA0827352 SGEA0827353 SGEA082737 SGEA0827375 558-0510 SGEA082740 SGEA082742 SGEA082745 SGEA082747 SGEA082760 SGEA082767 SGEA082768 SGEA082770 SGEA082708 SGEA082710 SGEA08277028 SGEA0827075 SGEA0827120 SGEA0827615
Glass lined tubing, GLT™
Slangen Chromatography Tubing
GLT™ (Glass Lined Tubing) is made by fusing a borosilicate glass lining onto the inside surface of stainless steel tubing. Ideal for HPLC columns, especially biological matrices, GC/MS interfaces, FTIR transfer lines, inlet liners, tee pieces and unions, thermal desorption tubes and SFC.
  • Excellent resistance to strong acids and bases
  • Stable and withstands high temperatures up to 500 °C
  • Can be bent without damage to the glass lining by heating
  • Graphite ferrules is recommended for connections
  • Used as straight tubing or it can be machined or shaped into virtually any chromatography accessory
Certificaten: RoHS compliant.